Assay Details
| Gene Symbol: AT3G18450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112730.4 | NP_188474.1 | 2 - 3 | 413 | 89 | |
| GenBank mRNA | BT010855.1 | - | 2 - 3 | 372 | 89 | |
| BT011321.1 | - | 2 - 3 | 272 | 89 | ||
More Information
Set Membership: |
|

AT3G18450
Genomic Map