Assay Details
| Gene Symbol: AT3G18450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112730.4 | NP_188474.1 | 3 - 4 | 624 | 65 | |
| GenBank mRNA | BT010855.1 | - | 3 - 4 | 583 | 65 | |
| BT011321.1 | - | 3 - 4 | 483 | 65 | ||
More Information
Set Membership: |
|

AT3G18450
Genomic Map