Assay Details
Gene Symbol: CASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112733.3 | NP_566611.1 | 10 - 11 | 1386 | 103 | |
GenBank mRNA | AY056225.1 | - | 10 - 11 | 1330 | 103 | |
AY142680.1 | - | 10 - 11 | 1154 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
