Assay Details
Gene Symbol: CASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112733.3 | NP_566611.1 | 16 - 17 | 2075 | 85 | |
GenBank mRNA | AY056225.1 | - | 16 - 17 | 2019 | 85 | |
AY142680.1 | - | 16 - 17 | 1843 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
