Assay Details
| Gene Symbol: CASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112733.3 | NP_566611.1 | 18 - 19 | 2236 | 76 | |
| GenBank mRNA | AY056225.1 | - | 18 - 19 | 2180 | 76 | |
| AY142680.1 | - | 18 - 19 | 2004 | 76 | ||
More Information
Set Membership: |
|

CASP
Genomic Map