Assay Details
| Gene Symbol: CASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112733.3 | NP_566611.1 | 2 - 3 | 447 | 137 | |
| GenBank mRNA | AY056225.1 | - | 2 - 3 | 391 | 137 | |
| AY142680.1 | - | 2 - 3 | 215 | 137 | ||
More Information
Set Membership: |
|

CASP
Genomic Map