Assay Details
| Gene Symbol: NUC-L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338347.1 | NP_001325664.1 | 9 - 10 | 1010 | 157 | |
| NM_001338348.1 | NP_001325663.1 | 10 - 11 | 1043 | 157 | ||
| NM_112747.3 | NP_188491.1 | 11 - 12 | 1075 | 157 | ||
| GenBank mRNA | DQ446672.1 | - | 11 - 12 | 1066 | 157 | |
More Information
Set Membership: |
|

NUC-L2
Genomic Map