Assay Details
| Gene Symbol: NUC-L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338347.1 | NP_001325664.1 | 13 - 14 | 1342 | 80 | |
| NM_001338348.1 | NP_001325663.1 | 14 - 15 | 1375 | 80 | ||
| NM_112747.3 | NP_188491.1 | 15 - 16 | 1407 | 80 | ||
| GenBank mRNA | DQ446672.1 | - | 15 - 16 | 1398 | 80 | |
More Information
Set Membership: |
|

NUC-L2
Genomic Map