Assay Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112768.3 | NP_188512.1 | 5 - 6 | 752 | 63 | |
GenBank mRNA | AB071846.1 | - | 5 - 6 | 554 | 63 | |
AY035137.1 | - | 5 - 6 | 641 | 63 | ||
AY059072.1 | - | 5 - 6 | 554 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
