Assay Details
Gene Symbol: LARP6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338382.1 | NP_001325674.1 | 7 - 8 | 1078 | 144 | |
NM_112796.2 | NP_188540.1 | 8 - 9 | 1159 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
