Assay Details
| Gene Symbol: PEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112829.5 | NP_188573.1 | 11 - 12 | 1599 | 144 | |
| GenBank mRNA | AY070042.1 | - | 11 - 12 | 1464 | 144 | |
More Information
Set Membership: |
|

PEN2
Genomic Map