Assay Details
| Gene Symbol: PEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112829.5 | NP_188573.1 | 5 - 6 | 1128 | 91 | |
| GenBank mRNA | AY070042.1 | - | 5 - 6 | 993 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PEN2| Gene Symbol: PEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112829.5 | NP_188573.1 | 5 - 6 | 1128 | 91 | |
| GenBank mRNA | AY070042.1 | - | 5 - 6 | 993 | 91 | |
Set Membership: |
|