Assay Details
Gene Symbol: BUB3.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112849.5 | NP_566644.1 | 1 - 2 | 293 | 68 | |
GenBank mRNA | AY087404.1 | - | 1 - 2 | 218 | 68 | |
BT004273.1 | - | 1 - 2 | 221 | 68 | ||
BT006152.1 | - | 1 - 2 | 117 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
