Assay Details
Gene Symbol: BUB3.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112849.5 | NP_566644.1 | 7 - 8 | 1005 | 88 | |
GenBank mRNA | AY087404.1 | - | 7 - 8 | 930 | 88 | |
BT004273.1 | - | 7 - 8 | 933 | 88 | ||
BT006152.1 | - | 7 - 8 | 829 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
