Assay Details
| Gene Symbol: AT3G20010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338440.1 | NP_001319598.1 | 3 - 4 | 1037 | 83 | |
| NM_001338443.1 | NP_001327761.1 | 3 - 4 | 1020 | 83 | ||
| NM_001338444.1 | NP_001327760.1 | 3 - 4 | 1022 | 83 | ||
| NM_001338445.1 | NP_001327759.1 | 2 - 3 | 783 | 83 | ||
| NM_001338446.1 | NP_001327756.1 | 2 - 3 | 783 | 83 | ||
| NM_112891.3 | NP_188635.1 | 2 - 3 | 783 | 83 | ||
More Information
Set Membership: |
|

AT3G20010
Genomic Map