Assay Details
| Gene Symbol: AT3G20010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338440.1 | NP_001319598.1 | 9 - 10 | 3088 | 73 | |
| NM_001338441.1 | NP_001327757.1 | 7 - 8 | 3690 | 73 | ||
| NM_001338442.1 | NP_001327758.1 | 8 - 9 | 3616 | 73 | ||
| NM_001338444.1 | NP_001327760.1 | 9 - 10 | 3073 | 73 | ||
| NM_112891.3 | NP_188635.1 | 8 - 9 | 2834 | 73 | ||
More Information
Set Membership: |
|

AT3G20010
Genomic Map