Assay Details
| Gene Symbol: TCP-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112896.4 | NP_188640.1 | 3 - 4 | 393 | 132 | |
| GenBank mRNA | BT021103.1 | - | 3 - 4 | 275 | 132 | |
| D11351.1 | - | 3 - 4 | 358 | 132 | ||
More Information
Set Membership: |
|

TCP-1
Genomic Map