Assay Details
| Gene Symbol: TCP-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112896.4 | NP_188640.1 | 4 - 5 | 481 | 120 | |
| GenBank mRNA | BT021103.1 | - | 4 - 5 | 363 | 120 | |
| D11351.1 | - | 4 - 5 | 446 | 120 | ||
More Information
Set Membership: |
|

TCP-1
Genomic Map