Assay Details
Gene Symbol: TCP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112896.4 | NP_188640.1 | 7 - 8 | 692 | 73 | |
GenBank mRNA | BT021103.1 | - | 7 - 8 | 574 | 73 | |
D11351.1 | - | 7 - 8 | 657 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
