Assay Details
| Gene Symbol: EHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035654.2 | NP_001030731.1 | 14 - 15 | 1432 | 117 | |
| NM_001338474.1 | NP_001327849.1 | 15 - 16 | 1510 | 117 | ||
| NM_112920.4 | NP_566657.1 | 15 - 16 | 1577 | 117 | ||
| GenBank mRNA | AK317308.1 | - | 14 - 15 | 1498 | 117 | |
| AY034911.1 | - | 15 - 16 | 1434 | 117 | ||
| BT004414.1 | - | 14 - 15 | 1304 | 117 | ||
More Information
Set Membership: |
|

EHD1
Genomic Map