Assay Details
| Gene Symbol: EHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035654.2 | NP_001030731.1 | 6 - 7 | 515 | 75 | |
| NM_001338474.1 | NP_001327849.1 | 7 - 8 | 593 | 75 | ||
| NM_112920.4 | NP_566657.1 | 7 - 8 | 660 | 75 | ||
| GenBank mRNA | AK317308.1 | - | 6 - 7 | 581 | 75 | |
| AY034911.1 | - | 7 - 8 | 517 | 75 | ||
| BT004414.1 | - | 6 - 7 | 387 | 75 | ||
More Information
Set Membership: |
|

EHD1
Genomic Map