Assay Details
Gene Symbol: AT3G20350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112926.5 | NP_188670.1 | 1 - 2 | 670 | 88 | |
GenBank mRNA | BT053761.1 | - | 1 - 2 | 378 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
