Assay Details
| Gene Symbol: PDIL5-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112948.3 | NP_566664.1 | 6 - 7 | 692 | 159 | |
| GenBank mRNA | AY050453.1 | - | 6 - 7 | 670 | 159 | |
More Information
Set Membership: |
|

PDIL5-3
Genomic Map