Assay Details
Gene Symbol: BSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119689.4 | NP_567980.1 | 5 - 6 | 1132 | 92 | |
GenBank mRNA | AF439824.1 | - | 5 - 6 | 865 | 92 | |
AY142063.1 | - | 5 - 6 | 815 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
