Assay Details
Gene Symbol: BSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119689.4 | NP_567980.1 | 6 - 7 | 1323 | 111 | |
GenBank mRNA | AF439824.1 | - | 6 - 7 | 1056 | 111 | |
AY142063.1 | - | 6 - 7 | 1006 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
