Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119692.3 | NP_195252.1 | 2 - 3 | 890 | 78 | |
| GenBank mRNA | AF428360.1 | - | 2 - 3 | 793 | 78 | |
| AY049260.1 | - | 2 - 3 | 793 | 78 | ||
| AY129494.1 | - | 2 - 3 | 752 | 78 | ||
More Information
Set Membership: |
|

IDH1
Genomic Map