Assay Details
| Gene Symbol: CLC-E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203995.1 | NP_001190924.1 | 1 - 2 | 714 | 98 | |
| NM_119709.1 | NP_567985.1 | 1 - 2 | 714 | 98 | ||
More Information
Set Membership: |
|

CLC-E
Genomic Map