Assay Details
| Gene Symbol: HMGB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342361.1 | NP_001329802.1 | 5 - 6 | 423 | 92 | |
| NM_119722.3 | NP_195282.1 | 6 - 7 | 414 | 92 | ||
| GenBank mRNA | AK317173.1 | - | 6 - 7 | 382 | 92 | |
| AY085615.1 | - | 6 - 7 | 386 | 92 | ||
| BT025597.1 | - | 5 - 6 | 321 | 92 | ||
| Y14075.1 | - | 321 | 92 | |||
More Information
Set Membership: |
|

HMGB5
Genomic Map