Assay Details
Gene Symbol: CST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203999.1 | NP_001190928.1 | 5 - 6 | 1058 | 88 | |
NM_119725.5 | NP_195285.3 | 5 - 6 | 1104 | 88 | ||
GenBank mRNA | AY065403.1 | - | 5 - 6 | 893 | 88 | |
AY096501.1 | - | 5 - 6 | 809 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
