Assay Details
| Gene Symbol: SERAT3;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119729.4 | NP_195289.3 | 3 - 4 | 888 | 111 | |
| GenBank mRNA | BT004080.1 | - | 3 - 4 | 562 | 111 | |
| BT005047.1 | - | 3 - 4 | 508 | 111 | ||
More Information
Set Membership: |
|

SERAT3;2
Genomic Map