Assay Details
| Gene Symbol: SERAT3;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119729.4 | NP_195289.3 | 8 - 9 | 1352 | 119 | |
| GenBank mRNA | BT004080.1 | - | 8 - 9 | 1026 | 119 | |
| BT005047.1 | - | 8 - 9 | 972 | 119 | ||
More Information
Set Membership: |
|

SERAT3;2
Genomic Map