Assay Details
Gene Symbol: LARP1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342378.1 | NP_001328621.1 | 1 - 2 | 862 | 67 | |
NM_119755.3 | NP_567991.1 | 1 - 2 | 862 | 67 | ||
GenBank mRNA | AF370207.1 | - | 1 - 2 | 813 | 67 | |
AY133869.1 | - | 1 - 2 | 520 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
