Assay Details
| Gene Symbol: AT4G36520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119815.7 | NP_195370.5 | 7 - 8 | 4374 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G36520| Gene Symbol: AT4G36520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119815.7 | NP_195370.5 | 7 - 8 | 4374 | 93 | |
Set Membership: |
|