Assay Details
Gene Symbol: APP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342416.1 | NP_001328936.1 | 10 - 11 | 1483 | 78 | |
NM_001342417.1 | NP_001328935.1 | 10 - 11 | 1483 | 78 | ||
NM_119840.7 | NP_195394.4 | 10 - 11 | 1563 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
