Assay Details
Gene Symbol: APP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342416.1 | NP_001328936.1 | 7 - 8 | 1200 | 71 | |
NM_001342417.1 | NP_001328935.1 | 7 - 8 | 1200 | 71 | ||
NM_119840.7 | NP_195394.4 | 7 - 8 | 1280 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
