Assay Details
| Gene Symbol: LEJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119855.4 | NP_195409.1 | 2 - 3 | 559 | 100 | |
| GenBank mRNA | AY085164.1 | - | 2 - 3 | 388 | 100 | |
| BT003835.1 | - | 2 - 3 | 374 | 100 | ||
| BT005190.1 | - | 2 - 3 | 298 | 100 | ||
More Information
Set Membership: |
|

LEJ2
Genomic Map