Assay Details
| Gene Symbol: LEJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119855.4 | NP_195409.1 | 6 - 7 | 838 | 76 | |
| GenBank mRNA | AY085164.1 | - | 6 - 7 | 667 | 76 | |
| BT003835.1 | - | 6 - 7 | 653 | 76 | ||
| BT005190.1 | - | 6 - 7 | 577 | 76 | ||
More Information
Set Membership: |
|

LEJ2
Genomic Map