Assay Details
| Gene Symbol: PLP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119868.3 | NP_195422.3 | 6 - 7 | 1352 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLP4| Gene Symbol: PLP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119868.3 | NP_195422.3 | 6 - 7 | 1352 | 93 | |
Set Membership: |
|