Assay Details
| Gene Symbol: SHM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119954.4 | NP_195506.1 | 9 - 10 | 1204 | 98 | |
| GenBank mRNA | AF428388.1 | - | 9 - 10 | 983 | 98 | |
| AJ271726.1 | - | 9 - 10 | 1028 | 98 | ||
| AY057645.1 | - | 9 - 10 | 982 | 98 | ||
| AY070726.1 | - | 9 - 10 | 1036 | 98 | ||
| BT006353.1 | - | 9 - 10 | 929 | 98 | ||
More Information
Set Membership: |
|

SHM1
Genomic Map