Assay Details
Gene Symbol: CAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342466.1 | NP_001327960.1 | 3 - 4 | 642 | 65 | |
NM_001342467.1 | NP_001327959.1 | 3 - 4 | 642 | 65 | ||
NM_001342468.1 | NP_001327958.1 | 3 - 4 | 532 | 65 | ||
NM_119958.4 | NP_195510.1 | 3 - 4 | 642 | 65 | ||
GenBank mRNA | AY302075.1 | - | 3 - 4 | 508 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
