Assay Details
Gene Symbol: CAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342468.1 | NP_001327958.1 | 4 - 5 | 754 | 100 | |
NM_119958.4 | NP_195510.1 | 4 - 5 | 864 | 100 | ||
GenBank mRNA | AY302075.1 | - | 4 - 5 | 730 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
