Assay Details
Gene Symbol: ELI3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119960.3 | NP_195512.1 | 1 - 2 | 410 | 118 | |
GenBank mRNA | AF361859.1 | - | 1 - 2 | 155 | 118 | |
AY129478.1 | - | 1 - 2 | 83 | 118 | ||
AY302080.1 | - | 1 - 2 | 83 | 118 | ||
X67815.1 | - | 128 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
