Assay Details
| Gene Symbol: FRS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119979.3 | NP_195531.1 | 1 - 2 | 2535 | 77 | |
| GenBank mRNA | AY136457.1 | - | 1 - 2 | 2469 | 77 | |
More Information
Set Membership: |
|

FRS5
Genomic Map