Assay Details
| Gene Symbol: AT4G38370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119999.3 | NP_568040.1 | 1 - 2 | 170 | 96 | |
| GenBank mRNA | AK226503.1 | - | 1 - 2 | 133 | 96 | |
More Information
Set Membership: |
|

AT4G38370
Genomic Map