Assay Details
| Gene Symbol: arpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202737.2 | NP_001189666.1 | 4 - 5 | 637 | 81 | |
| NM_001336451.1 | NP_001323477.1 | 4 - 5 | 637 | 81 | ||
| NM_001336452.1 | NP_001323478.1 | 4 - 5 | 635 | 81 | ||
| NM_179877.1 | NP_850208.1 | 4 - 5 | 504 | 81 | ||
More Information
Set Membership: |
|

arpc2b
Genomic Map