Assay Details
| Gene Symbol: arpc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202737.2 | NP_001189666.1 | 8 - 9 | 990 | 82 | |
| NM_001336452.1 | NP_001323478.1 | 8 - 9 | 988 | 82 | ||
| NM_179877.1 | NP_850208.1 | 8 - 9 | 830 | 82 | ||
More Information
Set Membership: |
|

arpc2b
Genomic Map