Assay Details
Gene Symbol: PHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179887.3 | NP_850218.1 | 3 - 4 | 2632 | 100 | |
GenBank mRNA | AY074292.1 | - | 3 - 4 | 2559 | 100 | |
AY091326.1 | - | 1 - 2 | 1407 | 100 | ||
DQ027037.1 | - | 1 - 2 | 1407 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
