Assay Details
Gene Symbol: PHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179887.3 | NP_850218.1 | 4 - 5 | 2767 | 103 | |
GenBank mRNA | AY074292.1 | - | 4 - 5 | 2694 | 103 | |
AY091326.1 | - | 2 - 3 | 1542 | 103 | ||
DQ027037.1 | - | 2 - 3 | 1542 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
