Assay Details
Gene Symbol: PHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179887.3 | NP_850218.1 | 5 - 6 | 3204 | 65 | |
GenBank mRNA | AY074292.1 | - | 5 - 6 | 3131 | 65 | |
AY091326.1 | - | 3 - 4 | 1979 | 65 | ||
DQ027037.1 | - | 3 - 4 | 1979 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
