Assay Details
| Gene Symbol: NF-YA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179904.5 | NP_850235.1 | 3 - 4 | 333 | 97 | |
| GenBank mRNA | AY072414.1 | - | 2 - 3 | 194 | 97 | |
| AY114711.1 | - | 2 - 3 | 141 | 97 | ||
More Information
Set Membership: |
|

NF-YA4
Genomic Map